Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium molybdate (di), Mo 56.5%

Catalog No. p-7048962
Click to view available options
Quantity:
50 g
250 g
1 kg

CAS: 27546-07-2 | H2MoO4 | 161.96 g/mol

Ammonium molybdate (di) is used as pigments, agricultural fertilizer, color lakes and fabric fire retardant agent. It is also used as a raw material for the production of molybdenum wire, ceramic glazes and other molybdenum compounds. Further, it is used in photography, ceramics and paints. It is also used reagent for analysis as well as in the determination of phosphorus in analytical chemistry. In addition, it is used as a catalyst in the petrochemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27546-07-2
Molecular Formula H2MoO4
Molecular Weight (g/mol) 161.96
MDL Number MFCD00010882
InChI Key VLAPMBHFAWRUQP-UHFFFAOYSA-L
Synonym Ammonium dimolybdate

Specifications

Physical Form Powder
Quantity 50 g
Solubility Information Soluble in water.
Formula Weight 339.95
Concentration or Composition (by Analyte or Components) Mo 56.5%
Chemical Name or Material Ammonium molybdate (di)
EINECSNumber 248-517-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.