Learn More
Ammonium molybdate (para) hydrate, Puratronic™, 99.999% (metals basis excluding W), W 300ppm max
Description
It is used for ceramics, decorating, electroplating, fixing of metals, fertilizers, photography, and as a negative stain in electron microscopy. It is also used in the manufacture of molybdenum. It is a useful reagent for detecting and determining phosphates, arsenates, lead and some alkaloids. It is useful in the preparation of catalysts for desulfurization and dehydrogenation reactions. Ammonium molybdate has been reported to control polycondensation reactions and decrease shrinkage values of metakaolin-based geopolymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | ∽190°C (decomposition) |
Physical Form | Powder |
Quantity | 5 g |
Assay Percent Range | 99.999% (Metal basis) |
Solubility Information | Freely soluble in water. |
Formula Weight | 1163.79 (Anhydrous) |
Appearance | Powder |
Density | 2.498 g/cm3 |
Chemical Name or Material | Ammonium molybdate (para) hydrate |
Safety and Handling
Hazard Category | H302 |
Precautionary Statement | P264b-P270-P301+P312-P330-P501c |
EINECSNumber | 234-722-4 |
RTECSNumber | QA4900000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.