Ammonium molybdate (para) tetrahydrate, ACS, 81-83% as MoO3, Alfa Aesar

Manufacturer: Alfa Aesar 1183136

Quantity 500g

 View more versions of this product

Catalog No. AA1183136

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12054-85-2
    Physical Form Powder
    Assay Percent Range 81-83%
    Molecular Formula H24Mo7N6O24·4H2O
    Formula Weight 1235.86
    MDL Number MFCD00167059
    Synonym Ammonium paramolybdate
    Chemical Name or Material Ammonium molybdate (para) tetrahydrate, (para) tetrahydrate
    Grade ACS Grade

    Specifications

    Formula Weight 1235.86
    Quantity 500g
    EINECS Number 234-722-4
    Physical Form Powder
    Assay Percent Range 81-83%
    Molecular Formula H24Mo7N6O24·4H2O
    MDL Number MFCD00167059
    CAS 12054-85-2
    Synonym Ammonium paramolybdate
    Health Hazard 1 H315-H319-H335
    Odor Odorless
    Appearance Powder
    Chemical Name or Material Ammonium molybdate (para) tetrahydrate, (para) tetrahydrate
    Grade ACS Grade

    H315-H319-H335

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit