Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium Molybdate Reagent I for Phosphorus by the Stannous Chloride Method (without extraction), Reagents
Ammonium Molybdate Reagent I for Phosphorus by the Stannous Chloride Method (without extraction), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc CA220000500A
| Quantity | 500 mL |
|---|
Chemical Identifiers
| 7732-18-5, 7664-93-9, 7664-93-9 | |
| 18.02 | |
| water |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| O |
Specifications
| Colorless to Light Yellow | |
| H2O | |
| O | |
| 18.02 | |
| Poly Bottle | |
| <3 | |
| Non-sterile | |
| Ammonium Molybdate Reagent I for Phosphorus by the Stannous Chloride Method (without extraction) |
| 7732-18-5, 7664-93-9, 7664-93-9 | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| Reagent | |
| 1.3 | |
| 500 mL | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction