Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Molybdate Solution, 48 g/L, Ricca Chemical
SDP

Catalog No. 665816 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
665-816 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 665-816 Supplier Ricca Chemical Company Supplier No. 665.816
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 12054-85-2 , 7732-18-5 , 1336-21-6
Molecular Formula H6MoNO2
Molecular Weight (g/mol) 148.00
InChI Key XCOMAVCHADVANQ-UHFFFAOYSA-O
Synonym Molybdic acid ammonium salt tetrahydrate, Hexammonium heptamolybdate 4-hydrate
PubChem CID 3084102
IUPAC Name ammonium hydrate molybdenum oxidandiide
SMILES [NH4+].O.[O--].[Mo]

Specifications

Color Colorless to Green-Yellow
CAS Min % 4.48, 93.0, 0.21
CAS Max % 4.66, 97.0, 0.22
Grade Laboratory
Packaging Natural Poly Bottle
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 48g/L
Chemical Name or Material Ammonium Molybdate Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.