Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium Molybdate Tetrahydrate, ACS Reagent, Reagents
Ammonium Molybdate Tetrahydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc C11350003A5
Quantity | 3 kg |
---|
Chemical Identifiers
12054-85-2 | |
148.00 | |
Ammonium molybdenum oxide | |
[NH4+].O.[O--].[Mo] |
H6MoNO2 | |
XCOMAVCHADVANQ-UHFFFAOYSA-O | |
ammonium hydrate molybdenum oxidandiide |
Specifications
White to Slightly Yellow-Green | |
190°C | |
H6MoNO2 | |
XCOMAVCHADVANQ-UHFFFAOYSA-O | |
ammonium hydrate molybdenum oxidandiide | |
ACS | |
2.498 | |
Non-sterile | |
Ammonium Molybdate Tetrahydrate, ACS Reagent |
12054-85-2 | |
190°C | |
Ammonium molybdenum oxide | |
[NH4+].O.[O--].[Mo] | |
148.00 | |
Poly Bottle | |
3 kg | |
Solid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction