Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium Molybdate TS, Ricca Chemical

Click to view available options
Quantity:
120 mL
500 mL
Specifications
Specifications
CAS Min % | 44.0, 38.74, 10.51, 5.24 |
CAS Max % | 45.0, 40.32, 10.94, 5.45 |
Grade | USP |
Packaging | Amber Poly Bottle |
Quantity | 500 mL |
Chemical Name or Material | AMMONIUM MOLYBDATE |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction