Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium molybdate(VI) tetrahydrate, 99+%, for analysis, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
12054-85-2
H24Mo7N6O24·4H2O
1235.86
12054-85-2
H24Mo7N6O24·4H2O
1235.86
Oxidation reagent for analysis
Assay Percent Range 99% min
Linear Formula (NH4)6Mo7O24·4H2O
Merck Index 15,53
Solubility Information Solubility in water: 400g/L (20°C). Other solubilities: practically insoluble in alcohol
Formula Weight 1235.86
Physical Form Crystalline Powder
Percent Purity 99+%
Grade Analysis
Chloride 10ppm max
Chlorine (Cl) 10ppm max.
Density 2.4900g/mL
Heavy Metals (as Pb) 10ppm max.
Packaging Plastic bottle
Phosphate 5ppm max.
Solubility Solubility in water: 400g/L (20°C). Other solubilities: practically insoluble in alcohol
Specific Gravity 2.49
Sulfate 100ppm max.
Color White
Melting Point 190.0°C
Quantity 100g
Chemical Name or Material Ammonium molybdate(VI) tetrahydrate
Show More Show Less

GHS Signal Word: Warning