Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium nitrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog No. AA1063818
Click to view available options
Quantity:
5 g
50 g
250 g

It is used in agriculture as a high-nitrogen fertilizer. It is one of the main components of ammonium nitrate/fuel oil (ANFO). ANFO is used in coal mining, quarrying, metal mining, and civil construction. It is also used as an oxidizer in solid rocket propellants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6484-52-2
Molecular Formula H4N2O3
Molecular Weight (g/mol) 80.04
MDL Number MFCD00011425
InChI Key DVARTQFDIMZBAA-UHFFFAOYSA-O
Synonym ammonium nitrate, nitram, ammonium nitricum, ammonium saltpeter, nitrate of ammonia, nitric acid ammonium salt, nitrato amonico, herco prills, german saltpeter, merco prills
PubChem CID 22985
ChEBI CHEBI:63038
IUPAC Name ammonium nitrate
SMILES [NH4+].[O-][N+]([O-])=O

Specifications

Boiling Point 210°C (decomposition)
Melting Point 170°C
Color Colorless
Physical Form Crystalline
Quantity 50 g
Assay Percent Range 100.00%
Solubility Information Highly soluble in water; soluble in alcohol and liquid ammonia.
Formula Weight 80.04
Odor Odorless
Appearance Colorless crystalline
Density 1.72
Chemical Name or Material Ammonium nitrate
Show More Show Less
Hazard Category H272-H319
Precautionary Statement P220-P221-P264b-P271-P280-P280i-P305+P351+P338-P370+P380+P375-P372-P380-P501c
DOTInformation DOT Class: 5.1, Packaging Group: III
EINECSNumber 229-347-8
RTECSNumber BR9050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.