Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium nitrate, 99+%, for analysis
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
Specifications
Specifications
| Boiling Point | 210.0°C |
| Melting Point | 169.0°C |
| Color | White |
| pH | 4.5 to 6 (5% soln. at 20°C) |
| Physical Form | Adhering Crystals |
| Quantity | 1 kg |
| Assay Percent Range | 99+% |
| Linear Formula | NH4NO3 |
| Fieser | 01,39; 15,14 |
| Merck Index | 15,531 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement May intensify fire; oxidizer. Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Keep/Store away from clothing/ combustible materials. In case of fire: Use water spray for extinction. Wear protective gloves/protective clothing/ |
| EINECSNumber | 229-347-8 |
| RTECSNumber | BR9050000 |
| TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction