Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Nitrate, MilliporeSigma™

Catalog No. M1011881000 Shop All MilliporeSigma Products
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic Bottle

For analysis EMSURE™ ACS

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6484-52-2
Molecular Formula H4N2O3
Molecular Weight (g/mol) 80.04
MDL Number MFCD00011425
InChI Key DVARTQFDIMZBAA-UHFFFAOYSA-O
Synonym ammonium nitrate, nitram, ammonium nitricum, ammonium saltpeter, nitrate of ammonia, nitric acid ammonium salt, nitrato amonico, herco prills, german saltpeter, merco prills
PubChem CID 22985
ChEBI CHEBI:63038
IUPAC Name ammonium nitrate
SMILES [NH4+].[O-][N+]([O-])=O

Specifications

Boiling Point 210°C
Quantity 1 kg
UN Number UN1942
Solubility Information Soluble in water: 1920g/L at 20°C
Formula Weight 80.04
Odor Odorless
Packaging Plastic Bottle
Density 1.72g/cm3 at 20°C
Chemical Name or Material Ammonium nitrate
Hazard Category H272: May intensify fire; oxidiser.
DOTInformation DOT Class: 5.1, Packaging Group: III
EINECSNumber 229-347-8
RTECSNumber BR9050000
Recommended Storage Store at 5° to 30°C (41° to 86°F), Tightly closed. Dry. Protect from Light. Away from combustible material and sources of ignition and heat.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.