Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Nitrate, Granular, Reagent, ACS, 95%, Spectrum™ Chemical
SDP

Catalog No. 18600628
Click to view available options
Quantity:
25 g
500 g
2.5 kg
6 x 500 g
12 kg
25 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

A1215, 6484-52-2, NH4NO3

Spectrum™ Chemical Ammonium Nitrate, Granular, Reagent, ACS is a white crystalline solid which is highly soluble in water. It is most commonly used in agriculture as a nitrogen fertilizer. It is a main component in some explosives and also finds use in instant cold packs as hydrating the salt is an endothermic process. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 6484-52-2
Molecular Formula H4N2O3
Molecular Weight (g/mol) 80.04
MDL Number MFCD00011425
InChI Key DVARTQFDIMZBAA-UHFFFAOYSA-O
IUPAC Name ammonium nitrate
SMILES [NH4+].[O-][N+]([O-])=O

Specifications

pH 4.5-6.0
CAS Max % 100%
Quantity 4 x 2.5 kg
UN Number UN1942
Percent Purity 95%
Grade ACS
Packaging Poly Bottle
Residue after Ignition 0.01%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.