Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ammonium O,O'-diethyl dithiophosphate, typically 95%

Manufacturer:  Alfa Aesar™ 3013914

Quantity 25g

 View more versions of this product

Catalog No. AA3013914


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1068-22-0
203.25
Diethyl dithiophosphate ammonium salt
(CH3CH2O)2P(S)SNH4
MFCD00012635
Ammonium O,O'-diethyl dithiophosphate

Specifications

1068-22-0
White
203.25
MFCD00012635
Unpleasant
Ammonium O,O'-diethyl dithiophosphate
Crystalline
(CH3CH2O)2P(S)SNH4
165° to 175°C
25g
Diethyl dithiophosphate ammonium salt
95% (typically)

Safety and Handling

SDS & Safety Documents

RTECSNumber : BP8145000

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit