Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium O,O'-diethyl dithiophosphate, typically 95%

Catalog No. p-7049831
Click to view available options
Quantity:
5 g
25 g

CAS: 1068-22-0 | C4H14NO2PS2 | 203.255 g/mol

Diethyl dithiophosphate ammonium salt can be used in the extraction of arsenic. It is used as a source of the (C2H5O)2PS2- ligand in coordination chemistry and in analytical chemistry for determination of various ions. It can be obtained by the reaction of phosphorus pentasulfide with ethanol and ammonia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1068-22-0
Molecular Formula C4H14NO2PS2
Molecular Weight (g/mol) 203.255
MDL Number MFCD00012635
InChI Key HFRHTRKMBOQLLL-UHFFFAOYSA-N
Synonym diethyl dithiophosphate, ammonium salt, ammonium ethyl phosphorodithioate, diethyl dithiophosphate ammonium salt, ammonium o,o-diethyldithiophosphate, ammonium diethyl dithiophosphate, c4h11o2ps2.nh3, wln: shps&o2&o2 &zh, ammonium di-o-ethyl dithiophosphate, ammonium,o-diethyl phosphorodithioate, ammonium o,o'-diethyl dithiophosphate
PubChem CID 12614240
IUPAC Name azane;diethoxy-sulfanyl-sulfanylidene-$l^{5}-phosphane
SMILES CCOP(=S)(OCC)S.N

Specifications

Melting Point 165°C to 175°C
Color White
Odor Unpleasant
Linear Formula (CH3CH2O)2P(S)SNH4
Quantity 5 g
Solubility Information Soluble in water,methanol,ethanol,pyridine,acetone & ammonia. Insoluble in ethyl acetate,methyl chloride and chloroform.
Formula Weight 203.25
Percent Purity 95% (typically)
Physical Form Crystalline
Chemical Name or Material Ammonium O,O'-diethyl dithiophosphate
EINECSNumber 213-942-4
RTECSNumber BP8145000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.