Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium oxalate monohydrate, ACS, 99.0-101.0%

Catalog No. AA3622857
Click to view available options
Quantity:
100 g
500 g
4 x 500 g

In detinning of iron, metal polishing | CAS: 6009-70-7 | C2H10N2O5 | 142.111 g/mol

In detinning of iron, metal polishing, determination of lead, calcium, and rare earth metalsAmmonium oxalate monohydrate is widely utilized as a buffering reagent and a dispersant to determine the surface interaction of platelets. It finds an application to study

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • In detinning of iron, metal polishing, determination of lead, calcium, and rare earth metals
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6009-70-7
Molecular Formula C2H10N2O5
Molecular Weight (g/mol) 142.111
MDL Number MFCD00149694
InChI Key MSMNVXKYCPHLLN-UHFFFAOYSA-N
Synonym ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate
PubChem CID 516808
IUPAC Name diazanium;oxalate;hydrate
SMILES C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O

Specifications

Melting Point 70°C (decomposition)
Color White
Physical Form Granules
Quantity 4 x 500 g
Assay Percent Range 99.0-101.0%
Beilstein 3759830
Merck Index 14,537
Solubility Information Soluble in water; Slightly soluble in alcohol.
Formula Weight 142.11 (124.10 Anhydrous)
Percent Purity 99.0 to 101.0%
Grade ACS
Density 1.50 g/mL
Chemical Name or Material Ammonium oxalate monohydrate, Monohydrate
Show More Show Less
Hazard Category H302+H312-H320-H335-H372
Hazard Statement GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 214-202-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.