Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium Oxalate Monohydrate, Honeywell Fluka™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
250 g
500 g
1 kg
2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Quantity | 1 kg |
Linear Formula | (NH4)2C2O4 · H2O |
UN Number | NONH for all modes of transport |
Beilstein | 3759830 |
Formula Weight | 142.11g/mol |
Percent Purity | 99.5 to 101% |
Grade | ACS/puriss. p.a. |
Residue after Ignition | ≤0.02% (as SO4) |
Chemical Name or Material | Ammonium Oxalate Monohydrate, Reag. Ph. Eur. reag. ISO |
Safety and Handling
Hazard Statement | P301 + P312 + P330-P302 + P352 + P312 |
Precautionary Statement | H302 + H312 |
EINECSNumber | 238-135-4 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction