Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Oxalate Monohydrate, Solstice

Catalog No. 6002412 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
2.5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-024-12 2.5 kg
60-024-13 250 g
60-024-14 500 g
60-024-11 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-024-12 Supplier Solstice Supplier No. 323042.5KG

Puriss. p.a., ACS Reagent, Reagent ISO, Reagent European Pharmacopoeia, 99.5-101.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6009-70-7
Molecular Formula C2H10N2O5
Molecular Weight (g/mol) 142.111
MDL Number MFCD00149694
InChI Key MSMNVXKYCPHLLN-UHFFFAOYSA-N
Synonym ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate
PubChem CID 516808
IUPAC Name diazanium;oxalate;hydrate
SMILES C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O

Specifications

Quantity 2.5 kg
Linear Formula (NH4)2C2O4 · H2O
UN Number NONH for all modes of transport
Beilstein 3759830
Formula Weight 142.11g/mol
Percent Purity 99.5 to 101%
Grade ACS/puriss. p.a.
Residue after Ignition ≤0.02% (as SO4)
Chemical Name or Material Ammonium Oxalate Monohydrate, Reag. Ph. Eur. reag. ISO
Hazard Statement P301 + P312 + P330-P302 + P352 + P312
Precautionary Statement H302 + H312
EINECSNumber 238-135-4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.