Learn More
Ammonium peroxydisulfate, 98%
Description
Ammonium peroxydisulfate is widely utilized as an oxidizing agent and radical initiators in the polymerization of certain alkenes. It is actively involved in in the synthesis of commercially important polymers like styrene-butadiene rubber and polytetrafluoroethylene. It combines with tetramethylethylenediamine to catalyze the polymerization of acrylamide. Due to its oxidizing properties, it can be employed to etch copper on printed circuit boards which is an alternative to ferric chloride solution. Also, it acts as oxidants in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ammonium peroxydisulfate is widely utilized as an oxidizing agent and radical initiators in the polymerization of certain alkenes. It is actively involved in in the synthesis of commercially important polymers like styrene-butadiene rubber and polytetrafluoroethylene. It combines with tetramethylethylenediamine to catalyze the polymerization of acrylamide. Due to its oxidizing properties, it can be employed to etch copper on printed circuit boards which is an alternative to ferric chloride solution. Also, it acts as oxidants in organic synthesis.
Solubility
Freely soluble in water.
Notes
Aqueous solution decomposes slowly at room temperature.Moisture sensitive. Incompatible with strong reducing agents, organic materials and powdered metals.

Specifications
Specifications
Melting Point | 120°C (decomposition) |
Quantity | 5000 g |
Linear Formula | (NH4)2S2O8 |
UN Number | UN1444 |
Flash Point | None |
Merck Index | 14,541 |
Solubility Information | Freely soluble in water. |
Formula Weight | 228.19 |
Percent Purity | 98% |
Odor | Odorless |
Show More |
Safety and Handling
Hazard Category | H272-H302-H315-H317-H319-H334-H335 |
Hazard Statement | GHS H Statement H272-H301-H334-H315-H319-H317-H335 May intensify fire oxidizer. Toxic if swallowed. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation. |
Precautionary Statement | P210-P220-P221-P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P333+P313-P342+P311-P362-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: AMMONIUM PERSULFATE |
EINECSNumber | 231-786-5 |
RTECSNumber | SE0350000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.