Learn More
Ammonium peroxydisulfate, ACS, 98.0% min
Description
Ammonium peroxydisulfate is used as an oxidizer, a bleaching agent, in the field of photography, to make aniline dyes, electroplating and in analytical chemistry for determination of manganese.Ammonium peroxydisulfate is widely utilized as an oxidizing agent and radical initiators in the polymerization of certain alkenes. It is actively involved in in the synthesis of commercially important polymers like styrene-butadiene rubber and polytetrafluoroethylene. It combines with tetramethylethylenediamine to catalyze the polymerization of acrylamide. Due to its oxidizing properties, it can be employed to etch copper on printed circuit boards which is an alternative to ferric chloride solution. Also, it acts as oxidants in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 120°C (decomposition) |
Physical Form | Lump |
Quantity | 100 g |
Assay Percent Range | 98.0% min |
Flash Point | None |
Solubility Information | Freely soluble in water. |
Formula Weight | 228.19 |
Grade | ACS |
Refractive Index | 1.5 |
Sensitivity | Moisture sensitive |
Show More |
Safety and Handling
Hazard Category | H272-H302-H315-H317-H319-H334-H335 |
Precautionary Statement | P210-P220-P221-P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P333+P313-P342+P311-P362-P370+P378q-P501c |
DOTInformation | Hazard Class: 5.1; Packaging Group: III |
EINECSNumber | 231-786-5 |
RTECSNumber | SE0350000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.