Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Peroxydisulfate, Crystal, BiotechGrade, 98%, Spectrum™ Chemical
SDP

Catalog No. 18600635
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail

A1227, 7727-54-0, (NH4)2S2O8

Spectrum™ Chemical Ammonium Peroxydisulfate, Crystal, BiotechGrade this highly water soluble oxidizing agent is a catalyst for acrylamide gel polymerization when used in conjunction with Tetramethylethylenediamine. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.

Chemical Identifiers

CAS 7727-54-0
Molecular Formula H8N2O8S2
Molecular Weight (g/mol) 228.19
MDL Number MFCD00003390
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
IUPAC Name diammonium [(sulfonatoperoxy)sulfonyl]oxidanide
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O

Specifications

CAS Max % 100%
Quantity 25 g
UN Number UN1444
Percent Purity 98%
Grade Biotech
Packaging Poly Bottle
Residue after Ignition 0.05%
Appearance Colorless to white
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.