Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Peroxydisulfate (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. A6823 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
3 kg
10 kg
Packaging:
Poly Bottle
Poly Pail

Catalog No. A682-3

Chemical Identifiers

CAS 7727-54-0
Molecular Formula H8N2O8S2
Molecular Weight (g/mol) 228.19
MDL Number MFCD00003390
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
Synonym ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
PubChem CID 62648
IUPAC Name diammonium [(sulfonatoperoxy)sulfonyl]oxidanide
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O

Specifications

Melting Point 120°C
Color White
pH 3.2
Physical Form Powder/Solid
Quantity 3 kg
Assay Percent Range ≥98.0 %
Linear Formula (NH4)2S2O8
Formula Weight 228.19
Percent Purity ≥98.0%
Grade Certified ACS
Identification Pass Test
Packaging Poly Bottle
Vapor Pressure Negligible
Decomposition Information >120°C
Density 1.980g/cm³
Chemical Name or Material Ammonium Peroxydisulfate
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Causes eye, skin, and respiratory tract irritation. May cause allergic respiratory and skin reaction. Use personal protective equipment. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:2
Flammability:1
Instability:2
DOTInformation DOT Class 5.1, : Oxidizer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.