Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium perrhenate(VII), 99+% (metals basis)

Catalog No. p-7048754
Click to view available options
Quantity:
2 g
10 g
50 g

Catalog No. AA1141004

Supplier: Thermo Scientific Chemicals 01141004

Only null left
Add to Cart

CAS: 13598-65-7 | H4NO4Re | 268.24 g/mol

Ammonium perrhenate(VII) is used to synthesis the rehenium(III) organohydrazides which is used as radiopharmaceuticals and also used in the manufacturing of rhenium metal and its additives in superalloys. It is used as catalyst for the synthesis of platinum-rhenium catalysts which used in gasoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13598-65-7
Molecular Formula H4NO4Re
Molecular Weight (g/mol) 268.24
MDL Number MFCD00012377
InChI Key XPSXHXWHJAIQJR-UHFFFAOYSA-M
Synonym azanium oxido trioxo rhenium, ammonium tetraoxorhenate vii, acmc-20alsh, nh4reo4, rhenate reo41-, ammonium, t-4, ammonium tetraoxorhenate 1-, rhenate reo41-, ammonium 1:1 , t-4, ammonium perrhenate, nh4 reo4, rhenate reo41-, ammonium, beta-4, ammonium perrhenate vii , puratronic metals basis
PubChem CID 3084163
SMILES N.O[Re](=O)(=O)=O

Specifications

Melting Point (decomposition)
Color White
Physical Form Powder
Quantity 2 g
Linear Formula NH4ReO4
UN Number UN1479
Solubility Information Soluble in water.
IUPAC Name ammonium tetraoxorheniumuide
Formula Weight 268.24
Percent Purity ≥99%
Density 3.97 g/mL
Assay (metals basis)
Chemical Name or Material Ammonium perrhenate(VII)
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H272
May intensify fire
oxidizer.
Precautionary Statement GHS P Statement
P221-P210-P220-P280-P370+P378a-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
EINECSNumber 237-075-6
RTECSNumber VI0710000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.