Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraphenylphosphonium bromide, 99%

Catalog No. AC204020250
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle

CAS: 2751-90-8 | C24H20BrP | 419.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2751-90-8
Molecular Formula C24H20BrP
Molecular Weight (g/mol) 419.30
MDL Number MFCD00011915
InChI Key BRKFQVAOMSWFDU-UHFFFAOYSA-M
Synonym tetraphenylphosphonium bromide, phosphonium, tetraphenyl-, bromide, tetraphenylphosphoniumbromide, tetraphenylphosponium bromide, tetraphenylphosphanium bromide, phosphonium, tetraphenyl-, bromide 1:1, ph4pbr, bromotetraphenylphosphorane, acmc-1ce9k, tetraphenylphosphorus bromide
PubChem CID 2724163
IUPAC Name tetraphenylphosphanium;bromide
SMILES [Br-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 295.0°C to 299.0°C
Color White
Density 260g/mL
Flash Point 260°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Plastic bottle
Linear Formula (C6H5)4PBr
Quantity 25 g
Beilstein 16, II, 373
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in alcohols
Formula Weight 419.29
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Tetraphenylphosphonium bromide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 220-393-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.