Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Phosphate Dibasic, puriss., Solstice

Catalog No. 6001824 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
25 kg
6 x 1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-018-24 1 kg
60-044-538 6 x 1 kg
60-018-25 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-018-24 Supplier Solstice Supplier No. 042111KG
Add to Cart
Add to Cart

Puriss., meets analytical specification of NF, 99-102%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-28-0
Molecular Formula H9N2O4P
Molecular Weight (g/mol) 132.06
MDL Number MFCD00010891
InChI Key MNNHAPBLZZVQHP-UHFFFAOYSA-N
Synonym diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate
PubChem CID 24540
ChEBI CHEBI:63051
IUPAC Name diazanium;hydrogen phosphate
SMILES N.N.OP(O)(O)=O

Specifications

Quantity 1 kg
Linear Formula (NH4)2HPO4
UN Number NONH for all modes of transport
Formula Weight 132.06g/mol
Percent Purity 99 to 102%
Grade puriss.
Chemical Name or Material Ammonium Phosphate Dibasic, Meets analytical specification of NF
EINECSNumber 231-987-8
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.