Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium Sulfate, BiotechGrade, 99%, Spectrum™ Chemical

Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Description
Spectrum™ Chemical Ammonium Sulfate, BiotechGrade is an inorganic salt that is very soluble in water It is a widely used reagent in molecular biology and chromatography for the precipitation and fractionation of proteins, crystallization of proteins and protein-nucleic acid complexes. Spectrum offers highly pure reagents suitable for biochemical research and analysis.
Specifications
Specifications
pH | 5.0-6.0 |
CAS Max % | 100% |
Quantity | 500 g |
Percent Purity | 99% |
Grade | Biotech |
Packaging | Poly Bottle |
Residue after Ignition | 0.005% |
Appearance | White, crystallized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction