Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium sulfate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99%, Solstice

Catalog No. 6002303 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
4 x 5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-023-03 5 kg
60-023-02 1 kg
60-045-906 4 x 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-023-03 Supplier Solstice Supplier No. 311195KG
Add to Cart
Add to Cart

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
PubChem CID 6097028
ChEBI CHEBI:62946
IUPAC Name diazanium;sulfate
SMILES N.N.OS(O)(=O)=O

Specifications

Quantity 5 kg
Linear Formula (NH4)2SO4
UN Number NONH for all modes of transport
Formula Weight 132.14g/mol
Percent Purity ≥99%
Grade ACS/puriss. p.a.
Residue after Ignition ≤0.005% (as SO4)
Density 1.77g/mL at 25°C
Chemical Name or Material Ammonium Sulfate, Reag. Ph. Eur. reag. ISO
EINECSNumber 231-984-1
RTECSNumber BS4500000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.