Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Sulfide, Light Solution, Reagent, 20-24%, Spectrum™ Chemical
SDP

Catalog No. 18600674 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
6 x 500 mL
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
Poly-Coated Glass Bottle

A1250, 12135-76-1, (NH4)2S

Spectrum™ Chemical Ammonium Sulfide, Light Solution, Reagent , is an unstable salt with the formula (NH4)2S. It has a very strong odor, and the Reagent grade means it is used in the laboratory as an analytical testing agent. Common uses of AMMONIUM SULFIDE, LIGHT SOLUTION, REAGENT are industrial applications including photo developing, textile manufacturing and others.

Chemical Identifiers

CAS 12135-76-1, 7732-18-5
Molecular Formula H8N2S
Molecular Weight (g/mol) 68.14
MDL Number MFCD00010892
InChI Key UXXGWUQNRMPNKH-UHFFFAOYSA-N
IUPAC Name diamine sulfane
SMILES N.N.S

Specifications

CAS Max % 80%
Quantity 100 mL
UN Number UN2683
Percent Purity 20 to 24%
Grade Reagent
Packaging Amber Glass Bottle
Residue after Ignition 0.04%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.