Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium tellurate, 99.5%, Thermo Scientific Chemicals

Catalog No. p-7049482
Click to view available options
Quantity:
10 g
50 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13453-06-0
Molecular Formula H8N2O4Te
Molecular Weight (g/mol) 227.67
MDL Number MFCD00015964
InChI Key DRGYXGZFRXFMHF-UHFFFAOYSA-N
Synonym ammonium tellurate, diammonium tellurate, diammonium tetraoxotellurate, ammonium tellurate nh4 2teo4, telluric acid, diammonium salt, telluric acid h2teo4 , diammonium salt, ammonium tellurate vi nh4 2teo4, telluric acid h2teo4 , ammonium salt 1:2, ammonium tellurate vi 6ci, diazanium tellurate
PubChem CID 61608
IUPAC Name diazanium;tellurate
SMILES [NH4+].[NH4+].[O-][Te]([O-])(=O)=O

Specifications

Color White
Physical Form Powder
Quantity 10 g
UN Number UN3284
Solubility Information Soluble in water.
Formula Weight 227.67
Percent Purity 99.5%
Chemical Name or Material Ammonium tellurate
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TELLURIUM COMPOUND, N.O.S.
EINECSNumber 236-622-6
RTECSNumber WY2415000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.