Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium tetrachloropalladate(II), 98%, ACROS Organics™

Click to view available options
Quantity:
2g
10g
Packaging:
Glass bottle
13820-40-1
H8Cl4N2Pd
284.28
MFCD00010884
IRKVTUVFHGUMMN-UHFFFAOYSA-L
ammonium tetrachloropalladate ii,ammonium palladium chloride,diammonium tetrachloropalladate 2-,ammonium chloropalladite,bisammonium tetrachloropalladate 2-,diammonium tetrachloro palladite,ammonium tetrachloropalladate,ccris 7828,palladate 2-, tetrachloro-, diammonium, sp-4-1,ammoniumpalladiumchloride
61682
CHEBI:60149
diazanium;tetrachloropalladium(2-)
[NH4+].[NH4+].Cl[Pd-2](Cl)(Cl)Cl
13820-40-1
H8Cl4N2Pd
284.28
MFCD00010884
IRKVTUVFHGUMMN-UHFFFAOYSA-L
ammonium tetrachloropalladate ii,ammonium palladium chloride,diammonium tetrachloropalladate 2-,ammonium chloropalladite,bisammonium tetrachloropalladate 2-,diammonium tetrachloro palladite,ammonium tetrachloropalladate,ccris 7828,palladate 2-, tetrachloro-, diammonium, sp-4-1,ammoniumpalladiumchloride
61682
CHEBI:60149
diazanium;tetrachloropalladium(2-)
[NH4+].[NH4+].Cl[Pd-2](Cl)(Cl)Cl
Assay Percent Range 98%
Linear Formula (NH4)2PdCl4
Fieser 03,81; 01,778; 02,303; 04,379; 05,496; 06,442; 09,344; 10,297; 11,389; 12,367; 13,103; 15,245; 16,259; 17,255
Solubility Information Solubility in water: soluble
Formula Weight 284.28
Physical Form Powder
Percent Purity 98%
Density 2.1700g/mL
Packaging Glass bottle
Palladium (Pd) 36.5 % min.
Solubility Solubility in water: soluble.
Specific Gravity 2.17
Color Black
Quantity 2g
Chemical Name or Material Ammonium tetrachloropalladate(II)
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.

GHS Signal Word: Danger

EINECSNumber : 237-498-6