Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium tetrachloropalladate(II), 98%

Catalog No. p-3776984
Click to view available options
Quantity:
2 g
10 g
Packaging:
Glass bottle

CAS: 13820-40-1 | Cl4H8N2Pd | 284.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13820-40-1
Molecular Formula Cl4H8N2Pd
Molecular Weight (g/mol) 284.30
MDL Number MFCD00010884
InChI Key IRKVTUVFHGUMMN-UHFFFAOYSA-L
Synonym ammonium tetrachloropalladate ii, ammonium palladium chloride, diammonium tetrachloropalladate 2-, ammonium chloropalladite, bisammonium tetrachloropalladate 2-, diammonium tetrachloro palladite, ammonium tetrachloropalladate, ccris 7828, palladate 2-, tetrachloro-, diammonium, sp-4-1, ammoniumpalladiumchloride
PubChem CID 61682
ChEBI CHEBI:60149
SMILES [NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Pd++]

Specifications

Color Black
Physical Form Powder
Quantity 2 g
Assay Percent Range 98%
Linear Formula (NH4)2PdCl4
Fieser 03,81; 01,778; 02,303; 04,379; 05,496; 06,442; 09,344; 10,297; 11,389; 12,367; 13,103; 15,245; 16,259; 17,255
Solubility Information Solubility in water: soluble
IUPAC Name diazanium;tetrachloropalladium(2-)
Formula Weight 284.28
Percent Purity 98%
Packaging Glass bottle
Specific Gravity 2.17
Density 2.1700g/mL
Chemical Name or Material Ammonium tetrachloropalladate(II)
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
EINECSNumber 237-498-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.