Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium tetrachloroplatinate(II), 99.9% (metals basis), Pt 51% min

Catalog No. p-7048624
Click to view available options
Quantity:
1 g
5 g

CAS: 13820-41-2 | Cl4H8N2Pt | 372.96 g/mol

Ammonium tetrachloroplatinate(II) is used in spectral analysis standard. It is also used in the preparation of platinum sponge and platinum catalytic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13820-41-2
Molecular Formula Cl4H8N2Pt
Molecular Weight (g/mol) 372.96
MDL Number MFCD00010885
InChI Key QJIMNDWDOXTTBR-UHFFFAOYSA-L
Synonym ammonium tetrachloroplatinate ii, diammonium tetrachloroplatinate, ammonium platinous chloride, diammonium tetrachloroplatinate 2-, ammonium tetrachloroplatinate, bis ammonium tetrachloroplatinate 2-, ammonium chloroplatinate ii, unii-z5ny20a9s2, h8cl4n2pt, epitope id:151521
IUPAC Name platinum(2+) diammonium tetrachloride
SMILES [NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Pt++]

Specifications

Melting Point 145°C (decomposition)
Color Red
Physical Form Crystalline
Quantity 1 g
Linear Formula (NH4)2PtCl4
UN Number UN3288
Solubility Information Soluble in water.
Formula Weight 372.98
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Pt 51% min.
Odor Odorless
Density 2.936 g/mL
Assay (metals basis)
Chemical Name or Material Ammonium tetrachloroplatinate(II)
Show More Show Less
Hazard Category H301-H315-H317-H318-H334
Hazard Statement GHS H Statement
H301-H334-H318-H290-H315-H317
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May be corrosive to metals.
Causes skin irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P330-P333+P313-P342+P311-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 237-499-1
RTECSNumber TP1840000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.