Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Ammonium Thiocyanate
SCI_ED

Catalog No. S25177 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Lab Grade - Crystalline Powder
Reagent Grade - Crystalline Powder
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25177 100 g Reagent Grade - Crystalline Powder
S25158 500 g Lab Grade - Crystalline Powder
S25154 500 g Reagent Grade - Crystalline Powder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S25177
Analytical reagent

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 1762-95-4
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.117
InChI Key SOIFLUNRINLCBN-UHFFFAOYSA-N
Synonym ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
PubChem CID 15666
IUPAC Name azanium;thiocyanate
SMILES C(#N)[S-].[NH4+]

Specifications

Color White
Quantity 100 g
Assay Percent Range 100% w/w
Linear Formula NH4SCN
Formula Weight 76.12
Grade Reagent
Packaging Reagent Grade - Crystalline Powder
Density 1.3 g/cm3
Chemical Name or Material Ammonium Thiocyanate
Safety handling Check Alert storage code Red
ShelfLife 2 years
DOTInformation DOT Class 9, III, : Miscellaneous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.