Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium thiocyanate, 99+%, extra pure, Acros Organics

Click to view available options
Quantity:
500g
1kg
2.5kg
10kg
Packaging:
Plastic Drum
Plastic bottle
1762-95-4
CH4N2S
76.12
MFCD00011428
SOIFLUNRINLCBN-UHFFFAOYSA-N
ammonium thiocyanate,ammoniumrhodanid,thiocyanic acid, ammonium salt,ammonium isothiocyanate,ammonium rhodanate,weedazol tl,trans-aid,ammonium sulfocyanate,ammonium sulfocyanide,rhodanine, ammonium salt
15666
azanium;thiocyanate
C(#N)[S-].[NH4+]
1762-95-4
CH4N2S
76.12
MFCD00011428
SOIFLUNRINLCBN-UHFFFAOYSA-N
ammonium thiocyanate,ammoniumrhodanid,thiocyanic acid, ammonium salt,ammonium isothiocyanate,ammonium rhodanate,weedazol tl,trans-aid,ammonium sulfocyanate,ammonium sulfocyanide,rhodanine, ammonium salt
15666
azanium;thiocyanate
C(#N)[S-].[NH4+]
Assay Percent Range 99% min. (on dried basis) (Argentometry)
Linear Formula NH4SCN
Merck Index 15,558
Solubility Information Solubility in water: 1600g/L (20°C). Other solubilities: soluble in alcohol,acetone and ammonia,practically insoluble in chcl3 and ethyl acetate
Formula Weight 76.12
Physical Form Crystals
Percent Purity 99+%
Grade Extra Pure
Density 1.3000g/mL
Iron (Fe) 2ppm max.
Packaging Plastic bottle
Solubility Solubility in water: 1600g/L (20°C). Other solubilities: soluble in alcohol, acetone and ammonia, practically insoluble in chcl3 and ethyl acetate
Specific Gravity 1.3
Water 1% Max.
Color White
Melting Point 149.0°C
pH 4.0 to 6.0 (5% aq. soln.)
Quantity 500g
Chemical Name or Material Ammonium thiocyanate
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye damage.
Harmful to aquatic life with long lasting effects.
Contact with acids liberates very toxic gas.

GHS P Statement
Keep container tightly closed.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF

GHS Signal Word: Danger

EINECSNumber : 217-175-6