Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Thiocyanate, Solstice

Catalog No. 6002308 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
6 x 500 g
6 x 1 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-023-08 500 g
60-023-04 100 g
60-023-06 250 g
60-023-05 1 kg
60-045-908 6 x 500 g
60-045-907 6 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60-023-08 Supplier Solstice Supplier No. 31120500G
Add to Cart
Add to Cart

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99% (AT)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1762-95-4
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.117
MDL Number MFCD00011428
InChI Key SOIFLUNRINLCBN-UHFFFAOYSA-N
Synonym ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
PubChem CID 15666
IUPAC Name azanium;thiocyanate
SMILES C(#N)[S-].[NH4+]

Specifications

Quantity 500 g
Linear Formula NH4SCN
Beilstein 3595135
Formula Weight 76.12g/mol
Percent Purity ≥99% (AT)
Grade ACS/puriss. p.a.
Residue after Ignition ≤0.02% (as SO4)
Chemical Name or Material Ammonium Thiocyanate, Reag. Ph. Eur. reag. ISO
Hazard Statement P273-P280
Precautionary Statement H302 + H312 + H332-H412
EINECSNumber 217-175-6
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.