Learn More
Ammonium thiosulfate, 96%
Description
Ammonium thiosulfate finds application for leaching of gold and silver since it forms strong complexes. It is widely utilized as a photographic fixing agent, which acts quicklier than sodium thiosulfate fixers. It serves as a fertilizer. It acts as an additive to coal-waste mixtures to inhibit the formation of dangerous dioxins and furans. It is used as an analytical reagent in laboratories.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ammonium thiosulfate finds application for leaching of gold and silver since it forms strong complexes. It is widely utilized as a photographic fixing agent, which acts quicklier than sodium thiosulfate fixers. It serves as a fertilizer. It acts as an additive to coal-waste mixtures to inhibit the formation of dangerous dioxins and furans. It is used as an analytical reagent in laboratories.
Solubility
Soluble in water, slightly soluble in acetone. Insoluble in ethanol and diethyl ether.
Notes
Hygroscopic. Incompatible with strong oxidizing agents, acids, magnesium and aluminum.

Specifications
Specifications
Melting Point | ∼180°C (decomposition) |
pH | 6.0 to 6.5 |
Quantity | 500 g |
Merck Index | 14,562 |
Solubility Information | Soluble in water,slightly soluble in acetone. Insoluble in ethanol and diethyl ether. |
Formula Weight | 148.2 |
Percent Purity | ≥96% |
Sensitivity | Hygroscopic |
Density | 1.64 |
Chemical Name or Material | Ammonium thiosulfate |
Safety and Handling
EINECSNumber | 231-982-0 |
RTECSNumber | XN6465000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.