Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium tungsten oxide, 99.99+% (metals basis)

Catalog No. AA4458818
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4458818 50 g
AA4458809 10 g
AA4458830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4458818 Supplier Thermo Scientific Chemicals Supplier No. 04458818
Only null left
Add to Cart

CAS: 11140-77-5 | H40N10O41W12 | 3042.429 g/mol

Ammonium Paratungstate is used as a source for high-purity tungsten oxides, tungsten metal powders, carbides, or as a laboratory reagent. Other applications are in the fields of absorbent gels, coloring agent in the porcelain industry, and the catalyst industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11140-77-5
Molecular Formula H40N10O41W12
Molecular Weight (g/mol) 3042.429
MDL Number MFCD00015956
InChI Key LMAJGSPZJYEENT-UHFFFAOYSA-D
Synonym ammonium para tungstate hydrate, ammonium tungstate trace metals basis, ammonium paratungstate trace metals basis, dodecaoxooctacyclo 21.1.1.1 1 , 3 .1 3 ,?.1?,?.1 1 1 , 1 3 .1 1 ?, 1 ?.1 1 ?, 2 1 dodecatungstoxane-5,7,9,11,13,15,17,19,21,23-decol decaamine
PubChem CID 71306883
SMILES N.N.N.N.N.N.N.N.N.N.O[W]12(=O)O[W]3(=O)(O[W](=O)(O3)(O[W]4(=O)(O[W](=O)(O4)(O[W]5(=O)(O[W]6(=O)(O5)O[W]7(=O)(O6)O[W](=O)(O7)(O[W]8(=O)(O[W](=O)(O8)(O[W](=O)(O1)(O2)O)O)O)O)O)O)O)O)O

Specifications

Physical Form Powder
Quantity 50 g
Solubility Information Soluble in warm water
Formula Weight 283.91
Percent Purity ≥99.99%
Assay (metals basis)
Chemical Name or Material Ammonium tungsten oxide
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.