Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium vanadium oxide, 99% (metals basis)

Catalog No. AA11091A1
Click to view available options
Quantity:
250 g
1 kg

CAS: 7803-55-6 | H4NO3V | 116.98 g/mol

Ammonium vanadium oxide is used as a catalyst in organic synthesis, and as an analytical reagent in laboratories. It is employed in the preparation of vanadate. It acts as a sensor for the determination of N-acetyltransferase activity in humans. Further, it finds use in dyes, varnishes, indelible inks, drier for paints, and in photography. With copper carbonate, it has been shown to enhance the catalytic activities. It is an environmental-friendly catalyst to prepare alfa-hydroxyphosphonate derivatives in high yield through the reaction of aryl/heteroaryl aldehydes with triethylphosphonate derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7803-55-6
Molecular Formula H4NO3V
Molecular Weight (g/mol) 116.98
MDL Number MFCD00011430
InChI Key YBVKNHXQSRDWAA-UHFFFAOYSA-M
Synonym ammonium metavanadate, ammonium vanadate v, ammonium monovanadate, unii-fl85px638g, vanadate vo31-, ammonium, vanadate vo31-, ammonium 1:1, ammonium vanadium oxide, ammoniummetavanadate, vanadic acid, hvo3 , ammonium salt, ammonium meta-vanadate
PubChem CID 516859
SMILES N.O[V](=O)=O

Specifications

Melting Point 200°C (decomposition)
Color White
Physical Form Powder
Quantity 1 kg
Assay Percent Range 99% (Metal basis)
Solubility Information Slightly soluble in water. Soluble in dilute ammonia and diethanolamine. Insoluble in ether,and alcohol.
IUPAC Name azanium;oxido(dioxo)vanadium
Formula Weight 116.98
Appearance Off-white powder
Density 2.33
Chemical Name or Material Ammonium vanadium oxide
Show More Show Less
Hazard Category H301-H319-H332-H361d-H372
Precautionary Statement P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P310-P304+P340-P305+P351+P338-P308+P313-P330-P501c
DOTInformation DOT Class: 6.1, Packaging Group: II
EINECSNumber 232-261-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.