Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AMN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111545
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111545 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111545 Supplier Invitrogen™ Supplier No. PA5111545
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LTDGAVEAVL TYCPQIRILL FHGCPLITDH SREVLEQLVG PNKLKQVTWT VY Highest antigen sequence indentity to the following orthologs: Rat - 85%, Mouse - 79%.

AMN1 belongs to the AMN1 family. The exact function of AMN1 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AMN1
Applications Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AMN1
Gene Accession No. Q8IY45
Gene Alias 5830467E07Rik; A230083D09; Amn1; antagonist of mitotic exit network 1; antagonist of mitotic exit network 1 homolog; antagonist of mitotic exit network 1 homolog (S. cerevisiae); C730024G19Rik; Protein AMN1 homolog; RGD1308119; Unknown (protein for MGC:133966)
Gene Symbols AMN1
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human AMN1. Recombinant protein control fragment (Product #RP-107155).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 196394
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.