Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Amnionless/AMN Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298805PCP
Description
Amnionless/AMN Polyclonal antibody specifically detects Amnionless/AMN in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Amnionless/AMN | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
amnionless homolog (mouse), PRO1028, protein amnionless, UNQ513/PRO1028, visceral endoderm-specific type 1 transmembrane protein | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Amnionless/AMN. | |
0.1 mL | |
metabolism | |
81693 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction