Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Amnionless Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551458
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551458 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551458 Supplier Invitrogen™ Supplier No. PA551458
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VSKLWVPNTD FDVAANWSQN RTPCAGGAVE FPADKMVSVL VQEGHAVSDM LLPLDGELVL ASGAGFGVSD VGSHLDCGAG EPAVFRDSDR FSWHDPHLWR SGDEAPGLFF VDAERVPCRH DDVFFPPSAS FRVGLGPGAS PVRVRSISAL Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 67%.

The protein encoded by this gene is a type I transmembrane protein. It is thought to modulate bone morphogenetic protein (BMP) receptor function by serving as an accessory or coreceptor, and thus facilitates or hinders BMP binding. It is known that the mouse AMN gene is expressed in the extraembryonic visceral endoderm layer during gastrulation, but it is found to be mutated in amnionless mouse. The encoded protein has sequence similarity to short gastrulation (Sog) and procollagen IIA proteins in Drosophila.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Amnionless
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Amn
Gene Accession No. Q9BXJ7
Gene Alias 5033428N14Rik; Amn; amnion associated transmembrane protein; amnionless; amnionless homolog; AV002116; PRO1028; Protein amnionless; Soluble protein amnionless; type I transmembrane protein; UNQ513/PRO1028; visceral endoderm-specific type 1 transmembrane protein
Gene Symbols Amn
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Amnionless. Recombinant protein control fragment (Product #RP-88867).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81693
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.