Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Amnionless Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5107057
Description
Sequence homology: Cow: 93%; Dog: 86%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rat: 86%; Zebrafish: 85%.
The protein encoded by this gene is a type I transmembrane protein. It is thought to modulate bone morphogenetic protein (BMP) receptor function by serving as an accessory or coreceptor, and thus facilitates or hinders BMP binding. It is known that the mouse AMN gene is expressed in the extraembryonic visceral endoderm layer during gastrulation, but it is found to be mutated in amnionless mouse. The encoded protein has sequence similarity to short gastrulation (Sog) and procollagen IIA proteins in Drosophila.
Specifications
| Amnionless | |
| Polyclonal | |
| Unconjugated | |
| Amn | |
| 5033428N14Rik; Amn; amnion associated transmembrane protein; amnionless; amnionless homolog; AV002116; PRO1028; Protein amnionless; Soluble protein amnionless; type I transmembrane protein; UNQ513/PRO1028; visceral endoderm-specific type 1 transmembrane protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 81693 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q9BXJ7 | |
| Amn | |
| Synthetic peptide directed towards the N terminal region of human AMN. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction