Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amodiaquin Dihydrochloride Dihydrate 98.0+%, TCI America™
SDP

Catalog No. A313325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A313325G 25 g
A31335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A313325G Supplier TCI America Supplier No. A313325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6398-98-7
Molecular Formula C20H28Cl3N3O3
Molecular Weight (g/mol) 464.812
MDL Number MFCD00078857
InChI Key YVNAYSHNIILOJS-UHFFFAOYSA-N
Synonym amodiaquin dihydrochloride dihydrate, amodiaquine hcl, amodiaquine hydrochloride, amodiaquine dihydrochloride dihydrate, unii-k6pw2s574l, 4-7-chloroquinolin-4-yl amino-2-diethylamino methyl phenol dihydrochloride dihydrate, amodiaquine hydrochloride usp, cam-aq 1
PubChem CID 64646
ChEBI CHEBI:50652
IUPAC Name 4-[(7-chloroquinolin-4-yl)amino]-2-(diethylaminomethyl)phenol;dihydrate;dihydrochloride
SMILES CCN(CC)CC1=C(C=CC(=C1)NC2=C3C=CC(=CC3=NC=C2)Cl)O.O.O.Cl.Cl

Specifications

Color Yellow
Quantity 25 g
Formula Weight 428.78
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Amodiaquin Dihydrochloride Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.