Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Amodiaquine Antibody (6D10), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP105106JF549
Description
Amodiaquine Monoclonal antibody specifically detects Amodiaquine in Nonspecies specific samples. It is validated for ELISASpecifications
Amodiaquine | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Non-species specific | |
Purified |
ELISA | |
6D10 | |
ELISA | |
Amodiaquin;Amodiaquine USP24;Amodiaquine, ring closed | |
4-amino-2(diethylaminomethyl)phenol a preparation of a carrier protein | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction