Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amorolfine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. A21611G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21611G 1 g
A2161200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21611G Supplier TCI America Supplier No. A21611G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 78613-38-4
Molecular Formula C21H36ClNO
Molecular Weight (g/mol) 353.975
MDL Number MFCD00903738
InChI Key XZKWIPVTHGWDCF-KUZYQSSXSA-N
Synonym amorolfine hydrochloride, amorolfine hcl, loceryl, curanail, locetar, odenil, amorolfin, pekiron, amorolfine hydrochloride jan
PubChem CID 54259
ChEBI CHEBI:59649
IUPAC Name (2R,6S)-2,6-dimethyl-4-[2-methyl-3-[4-(2-methylbutan-2-yl)phenyl]propyl]morpholine;hydrochloride
SMILES CCC(C)(C)C1=CC=C(C=C1)CC(C)CN2CC(OC(C2)C)C.Cl

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 353.98
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Amorolfine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.