Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AMOT Antibody (OTI1A8), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272429AF594
Description
AMOT Monoclonal specifically detects AMOT in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
AMOT | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
AMOT | |
Full length recombinant protein of human AMOT (NP_573572) produced in HEK293T cells. | |
0.1 mL | |
154796.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI1A8 | |
Western Blot | |
angiomotin, angiomotin p80 isoform, KIAA1071angiomotin p130 isoform | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction