Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AMOT Antibody (OTI1A8), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272429B
Description
AMOT Monoclonal specifically detects AMOT in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| AMOT | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| AMOT | |
| Full length recombinant protein of human AMOT (NP_573572) produced in HEK293T cells. | |
| 0.1 mL | |
| 154796.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI1A8 | |
| Western Blot | |
| angiomotin, angiomotin p80 isoform, KIAA1071angiomotin p130 isoform | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction