Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AMOT Mouse anti-Human, Porcine, Clone: 3H7B3, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | AMOT |
---|---|
Clone | 3H7B3 |
Concentration | 0.87 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6271
![]() ![]() |
Proteintech Group Inc
601561IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody only recognizes p130.
This gene belongs to the motin family of angiostatin binding proteins characterized by conserved coiled-coil domains and C-terminal PDZ binding motifs. The encoded protein is expressed predomitly in endothelial cells of capillaries as well as larger vessels of the placenta where it may mediate the inhibitory effect of angiostatin on tube formation and the migration of endothelial cells toward growth factors during the formation of new blood vessels. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
AMOT | |
0.87 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
AMOT, angiomotin, KIAA1071 | |
AMOT | |
IgG2b | |
Protein A | |
Antibody |
3H7B3 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Pig | |
Q4VCS5 | |
100158213, 154796 | |
peptide | |
Primary | |
-20°C | |
AMOT |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title