Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AMOTL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542267
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542267 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542267 Supplier Invitrogen™ Supplier No. PA542267
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LTQEDPQMVY QSARQEPQGQ EHQVDNTVME KQVRSTQPQQ NNEELPTYEE Sequence homology: Cow: 93%; Dog: 79%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%.

The protein encoded by this gene is a peripheral membrane protein that is a component of tight junctions or TJs. TJs form an apical junctional structure and act to control paracellular permeability and maintain cell polarity. This protein is related to angiomotin, an angiostatin binding protein that regulates endothelial cell migration and capillary formation. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AMOTL1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene AMOTL1
Gene Accession No. Q8IY63
Gene Alias 2310010G08Rik; 2310067L22Rik; 4932416D09Rik; Amotl1; angiomotin like 1; angiomotin-like 1; angiomotin-like protein 1; JEAP; junction-enriched and associated protein; junction-enriched and -associated protein; mFLJ00155
Gene Symbols AMOTL1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human AMOTL1 (aa 144-193).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 154810
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.