Learn More
Invitrogen™ AMOTL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551533
Description
Immunogen sequence: SPSACYSPSS PVQVLEDSTY FSPDFQLYSG RHETSALTVE ATSSIREKVV EDPLCNFHSP NFLRISEVEM RGSEDAAAGT VLQRLIQEQL RYGTPTENMN LLAIQHQATG SAGPAHPTNN FSSTENLTQE DPQMVYQSAR QEPQGQEHQV Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 32%.
The protein encoded by this gene is a peripheral membrane protein that is a component of tight junctions or TJs. TJs form an apical junctional structure and act to control paracellular permeability and maintain cell polarity. This protein is related to angiomotin, an angiostatin binding protein that regulates endothelial cell migration and capillary formation. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
AMOTL1 | |
Polyclonal | |
Unconjugated | |
AMOTL1 | |
2310010G08Rik; 2310067L22Rik; 4932416D09Rik; Amotl1; angiomotin like 1; angiomotin-like 1; angiomotin-like protein 1; JEAP; junction-enriched and associated protein; junction-enriched and -associated protein; mFLJ00155 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
154810 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IY63 | |
AMOTL1 | |
Recombinant protein corresponding to Human AMOTL1. Recombinant protein control fragment (Product #RP-94762). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.