Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AMOTL2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550322

Catalog No. PA550322


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total AMOTL2 protein.

Regulates the translocation of phosphorylated SRC to peripheral cell-matrix adhesion sites. Required for proper architecture of actin filaments. Inhibits the Wnt/beta-catenin signaling pathway, probably by recruiting CTNNB1 to recycling endosomes and hence preventing its translocation to the nucleus. Participates in angiogenesis. May play a role in the polarity, proliferation and migration of endothelial cells. Selectively promotes FGF-induced MAPK activation through SRC.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

AMOTL2
Polyclonal
Unconjugated
AMOTL2
AMOTL2; angiomotin like 2; angiomotin-like 2; angiomotin-like protein 2; AW549739; C79691; KIAA0989; LCCP; Leman coiled-coil protein; MASCOT; mKIAA0989
Rabbit
Antigen Affinity Chromatography
RUO
51421
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.7 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q9Y2J4
AMOTL2
Fusion protein corresponding to residues near the C terminal of human angiomotin like 2.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.